Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007202.3 | NP_009133.2 | 8 | 1385 | ||
XM_006721432.3 | XP_006721495.1 | 8 | 1380 | |||
XM_017024071.1 | XP_016879560.1 | 8 | 1380 | |||
GenBank mRNA | AF037439.1 | - | 8 | 1345 | ||
AK308297.1 | - | 8 | 1232 | |||
AK312438.1 | - | 8 | 1338 | |||
BC017055.1 | - | 8 | 1341 | 3882941 | ||
DQ270159.1 | - | 8 | 1208 | |||
KU178546.1 | - | 8 | 1208 | |||
KU178547.1 | - | 8 | 1179 | |||
KU178548.1 | - | 7 | 1025 |