Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007227.3 | NP_009158.3 | 1 | 1203 | ||
GenBank mRNA | BC066879.1 | - | 1 | 1203 | 6971748 | |
BC066880.1 | - | 1 | 1203 | 6971750 | ||
BC067455.1 | - | 1 | 1203 | 6971749 | ||
EU883573.1 | - | 1 | 1087 |
Set Membership: |
G-protein coupled receptor |