Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281425.1 | NP_001268354.1 | 6 | 603 | ||
NM_001281426.1 | NP_001268355.1 | 7 | 718 | |||
NM_001281427.1 | NP_001268356.1 | 6 | 657 | |||
NM_007234.4 | NP_009165.1 | 7 | 687 | |||
NM_024348.3 | NP_077324.1 | 7 | 784 | |||
GenBank mRNA | AF082513.1 | - | 7 | 644 | ||
AK055199.1 | - | 2 | 2859 | |||
AK091661.1 | - | 5 | 3714 | |||
BC000319.2 | - | 7 | 735 | 2821720 | ||
BC003004.1 | - | 7 | 735 | 2821720 | ||
BC107697.1 | - | 7 | 656 | 6454606 | ||
BG715167.1 | - | 7 | 668 | |||
BI596981.1 | - | 7 | 671 | |||
BI837796.1 | - | 6 | 562 | |||
CN257020.1 | - | 5 | 439 |