Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNRG SIRNA Assays ›
Gene Symbol:
SYNRG
Gene Name:
synergin, gamma
Gene Aliases:
AP1GBP1, SYNG
Chromosome Location:
Chr.17: 37514797 - 37613637 on Build GRCh38
UniGene ID:
Hs.594647
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136358
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNRG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163544.2
NP_001157016.1
9
1004
NM_001163545.2
NP_001157017.1
9
1001
NM_001163546.2
NP_001157018.1
9
1004
NM_007247.5
NP_009178.3
10
1238
NM_080550.4
NP_542117.3
9
1004
NM_198882.2
NP_942583.1
9
1004
XM_005256980.4
XP_005257037.1
11
1599
XM_017024076.1
XP_016879565.1
11
1599
XM_017024077.1
XP_016879566.1
11
1518
XM_017024078.1
XP_016879567.1
11
1599
XM_017024079.1
XP_016879568.1
11
1599
XM_017024080.1
XP_016879569.1
11
5651
XM_017024081.1
XP_016879570.1
11
5648
XM_017024082.1
XP_016879571.1
11
1599
XM_017024083.1
XP_016879572.1
11
1599
XM_017024084.1
XP_016879573.1
11
1599
XM_017024085.1
XP_016879574.1
11
1599
XM_017024086.1
XP_016879575.1
11
1599
XM_017024087.1
XP_016879576.1
11
1599
XM_017024088.1
XP_016879577.1
10
1365
XM_017024089.1
XP_016879578.1
11
1599
XM_017024090.1
XP_016879579.1
10
1296
XM_017024091.1
XP_016879580.1
10
1215
XM_017024092.1
XP_016879581.1
11
1599
XM_017024093.1
XP_016879582.1
10
1296
XM_017024094.1
XP_016879583.1
10
1215
XM_017024095.1
XP_016879584.1
10
1296
XM_017024096.1
XP_016879585.1
11
1599
XM_017024097.1
XP_016879586.1
10
1296
XM_017024098.1
XP_016879587.1
10
1296
XM_017024099.1
XP_016879588.1
9
981
XM_017024100.1
XP_016879589.1
10
1296
XM_017024101.1
XP_016879590.1
10
1215
XM_017024102.1
XP_016879591.1
9
981
XM_017024103.1
XP_016879592.1
9
1062
XM_017024104.1
XP_016879593.1
9
1062
XM_017024105.1
XP_016879594.1
11
1599
XM_017024106.1
XP_016879595.1
9
1062
GenBank mRNA
AF169548.1
-
10
1198
AK025335.1
-
3
394
AK126988.1
-
10
1966
AK308292.1
-
4
617
BC090930.1
-
9
955
3077104
BC117313.1
-
9
944
40125864
BC143478.1
-
9
947
9051986
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
endocytosis
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0