Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145047.1 | NP_001138519.1 | 7 | 432 | ||
NM_001145048.1 | NP_001138520.1 | 6 | 364 | |||
NM_001145049.1 | NP_001138521.1 | 4 | 257 | |||
NM_007266.3 | NP_009197.2 | 6 | 471 | |||
NR_026735.1 | 4 | 364 | ||||
GenBank mRNA | AB044661.1 | - | 6 | 432 | ||
AJ010842.1 | - | 6 | 361 | |||
AK298827.1 | - | 6 | 765 | |||
AK298866.1 | - | 6 | 471 | |||
AK302131.1 | - | 4 | 330 | |||
AK310340.1 | - | 7 | 422 | |||
AK311423.1 | - | 4 | 387 | |||
AK312992.1 | - | 6 | 443 | |||
AK316404.1 | - | 4 | 257 | |||
BC007451.2 | - | 6 | 415 | 3029376 |