Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284341.1 | NP_001271270.1 | 7 | 1243 | ||
NM_001284343.1 | NP_001271272.1 | 3 | 321 | |||
NM_007270.4 | NP_009201.2 | 6 | 1084 | |||
XM_011515115.2 | XP_011513417.1 | 6 | 1084 | |||
GenBank mRNA | AF089745.1 | - | 6 | 755 | ||
AK075331.1 | - | 6 | 1048 | |||
AK128597.1 | - | 7 | 1025 | |||
AK293440.1 | - | 3 | 299 | |||
AK293954.1 | - | 5 | 657 | |||
AK294266.1 | - | 5 | 868 | |||
AK300328.1 | - | 7 | 1206 | |||
BC101723.1 | - | 6 | 1012 | 8069229 | ||
BC112053.1 | - | 6 | 1012 | 8327521 | ||
BQ688269.1 | - | 3 | 321 |
Gene Symbol: FKBP9P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003949.1 | 2 | 63 | |||
GenBank mRNA | BM041005.1 | - | 2 | 64 |