Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007282.4 | NP_009213.1 | 11 | 2615 | ||
NM_183381.2 | NP_899237.1 | 10 | 2513 | |||
XM_005247092.3 | XP_005247149.1 | 10 | 2472 | |||
XM_011512373.2 | XP_011510675.1 | 12 | 2620 | |||
XM_011512374.2 | XP_011510676.1 | 11 | 2518 | |||
XM_011512376.2 | XP_011510678.1 | 10 | 1923 | |||
XM_017005654.1 | XP_016861143.1 | 11 | 2694 | |||
XM_017005655.1 | XP_016861144.1 | 10 | 2595 | |||
XM_017005656.1 | XP_016861145.1 | 11 | 2574 | |||
XM_017005657.1 | XP_016861146.1 | 11 | 2707 | |||
XM_017005658.1 | XP_016861147.1 | 10 | 2605 | |||
XM_017005659.1 | XP_016861148.1 | 9 | 2342 | |||
XM_017005660.1 | XP_016861149.1 | 9 | 2332 | |||
XM_017005661.1 | XP_016861150.1 | 4 | 1697 | |||
GenBank mRNA | AF037204.1 | - | 10 | 1982 | ||
AK090771.1 | - | 7 | 1603 | |||
BC009803.1 | - | 11 | 2094 | 4274992 |