Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP4E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252127.1 | NP_001239056.1 | 14 | 1731 | ||
NM_007347.4 | NP_031373.2 | 14 | 1708 | |||
XM_005254264.3 | XP_005254321.1 | 14 | 1526 | |||
XM_006720447.3 | XP_006720510.1 | 14 | 1886 | |||
XM_017022042.1 | XP_016877531.1 | 14 | 1741 | |||
GenBank mRNA | AB030653.1 | - | 14 | 1631 | ||
AF155156.2 | - | 14 | 1634 | |||
BC126308.1 | - | 14 | 1707 | 8992024 | ||
BC130466.1 | - | 14 | 1707 | 40146497 | ||
CR749604.1 | - | 14 | 1608 |