Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007349.3 | NP_031375.3 | 21 | 3588 | ||
XM_005249539.1 | XP_005249596.1 | 22 | 3727 | |||
XM_011515982.2 | XP_011514284.1 | 21 | 3702 | |||
XM_011515983.1 | XP_011514285.1 | 21 | 3588 | |||
XM_017011881.1 | XP_016867370.1 | 22 | 4900 | |||
GenBank mRNA | AK093584.1 | - | 15 | 2281 | ||
AK123600.1 | - | 22 | 3727 | |||
AK126674.1 | - | 13 | 3667 | |||
BC033781.1 | - | 16 | 2829 | 4509264 | ||
BX538201.1 | - | 15 | 2364 | |||
BX640616.1 | - | 21 | 5486 | |||
U80735.1 | - | 17 | 2571 |