Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007372.3 | NP_031398.2 | 5, 6 | 712 | ||
NM_203499.2 | NP_987095.1 | 4, 5 | 641 | |||
XM_006721657.1 | XP_006721720.1 | 5, 6 | 849 | |||
XM_017024111.1 | XP_016879600.1 | 6, 7 | 948 | |||
GenBank mRNA | AB036090.1 | - | 3, 4 | 437 | ||
AF083255.1 | - | 2, 3 | 97 | |||
AK095772.1 | - | 2, 3 | 99 | |||
AK292908.1 | - | 2, 3 | 224 | |||
AL527653.3 | - | 5, 6 | 706 | |||
AL545851.3 | - | 4, 5 | 609 | |||
BC015505.1 | - | 3, 4 | 422 | 3905607 | ||
BC078667.1 | - | 3, 4 | 380 | |||
BC093081.1 | - | 5, 6 | 676 | 30407786 |