Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007372.3 | NP_031398.2 | 8 | 973 | ||
NM_203499.2 | NP_987095.1 | 7 | 902 | |||
XM_006721657.1 | XP_006721720.1 | 8 | 1110 | |||
XM_011524259.2 | XP_011522561.1 | 2 | 911 | |||
XM_017024111.1 | XP_016879600.1 | 9 | 1209 | |||
XM_017024112.1 | XP_016879601.1 | 3 | 289 | |||
GenBank mRNA | AB036090.1 | - | 6 | 698 | ||
AF083255.1 | - | 5 | 358 | |||
AK095772.1 | - | 5 | 387 | |||
AK292908.1 | - | 5 | 485 | |||
BC015505.1 | - | 6 | 683 | 3905607 | ||
BC078667.1 | - | 6 | 641 | |||
BC093081.1 | - | 8 | 937 | 30407786 |