Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306153.1 | NP_001293082.1 | 2, 3 | 188 | ||
NM_001306156.1 | NP_001293085.1 | 2, 3 | 420 | |||
NM_012073.4 | NP_036205.1 | 2, 3 | 588 | |||
GenBank mRNA | AK289353.1 | - | 2, 3 | 186 | ||
AK293341.1 | - | 2, 3 | 257 | |||
AK301760.1 | - | 2, 3 | 180 | |||
AK301985.1 | - | 2, 3 | 227 | |||
AK303086.1 | - | 2, 3 | 217 | |||
AK316376.1 | - | 2, 3 | 175 | |||
BC002971.1 | - | 2, 3 | 163 | |||
BC006543.2 | - | 2, 3 | 203 | 2824199 | ||
BC035499.2 | - | 2, 3 | 250 | 4329194 | ||
BC040027.1 | - | 2, 3 | 220 | |||
BU858349.1 | - | 2, 3 | 356 | |||
D43950.2 | - | 2, 3 | 205 | |||
DB449481.1 | - | 421 | ||||
DC395133.1 | - | 2, 3 | 180 | |||
FJ224303.1 | - | 2, 3 | 339 |