Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNDBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012142.4 | NP_036274.3 | 3 | 442 | ||
NR_027514.2 | 3 | 442 | ||||
GenBank mRNA | AF082569.1 | - | 3 | 301 | ||
AF087852.1 | - | 3 | 287 | |||
AF113535.1 | - | 1 | 251 | |||
AF132034.1 | - | 3 | 257 | |||
AF246144.1 | - | 3 | 259 | |||
AK075296.1 | - | 1 | 256 | |||
AK225143.1 | - | 3 | 257 | |||
AK225645.1 | - | 3 | 271 | |||
AK290665.1 | - | 3 | 271 | |||
AK295017.1 | - | 3 | 271 | |||
AK308817.1 | - | 2 | 866 | |||
AL552334.3 | - | 3 | 381 | |||
BC009689.1 | - | 3 | 285 | 3896421 | ||
CR450331.1 | - | 3 | 211 | |||
DA506028.1 | - | 3 | 431 | |||
DB492361.1 | - | 3 | 440 |