Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNDBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012142.4 | NP_036274.3 | 8 | 947 | ||
NR_027513.2 | 6 | 770 | ||||
NR_027514.2 | 7 | 850 | ||||
NR_045998.1 | 7 | 867 | ||||
NR_045999.1 | 5 | 710 | ||||
XM_006720448.1 | XP_006720511.1 | 6 | 869 | |||
GenBank mRNA | AF082569.1 | - | 8 | 806 | ||
AF087852.1 | - | 8 | 792 | |||
AF113535.1 | - | 6 | 756 | |||
AF132034.1 | - | 8 | 762 | |||
AF246144.1 | - | 8 | 764 | |||
AK075146.1 | - | 8 | 776 | |||
AK075296.1 | - | 6 | 761 | |||
AK124193.1 | - | 3 | 1351 | |||
AK128849.1 | - | 4 | 1991 | |||
AK225143.1 | - | 8 | 762 | |||
AK225645.1 | - | 8 | 776 | |||
AK290665.1 | - | 8 | 776 | |||
AK290707.1 | - | 7 | 729 | |||
AK295017.1 | - | 7 | 679 | |||
AL552334.3 | - | 8 | 885 | |||
BC009689.1 | - | 8 | 790 | 3896421 | ||
BQ439439.1 | - | 5 | 524 | |||
CR450331.1 | - | 8 | 716 |