Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098484.2 | NP_001091954.1 | 4 | 488 | ||
NM_001134742.1 | NP_001128214.1 | 4 | 488 | |||
NM_003759.3 | NP_003750.1 | 1 | 435 | |||
XM_017008792.1 | XP_016864281.1 | 2 | 161 | |||
GenBank mRNA | AB470072.1 | - | 3 | 378 | ||
AF004813.1 | - | 2 | 311 | |||
AF007216.1 | - | 1 | 388 | |||
AF011390.1 | - | 4 | 488 | |||
AF053753.1 | - | 3 | 371 | |||
AF053754.1 | - | 3 | 371 | |||
AF069510.1 | - | 3 | 415 | |||
AF157492.1 | - | 4 | 447 | |||
AF310248.1 | - | 3 | 371 | |||
AK226164.1 | - | 2 | 261 | |||
AK307594.1 | - | 4 | 468 | |||
BC030977.1 | - | 1 | 435 | 4610968 | ||
BC127908.1 | - | 1 | 32 | |||
DA635432.1 | - | 1 | 444 | |||
EF531618.1 | - | 3 | 371 |