Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004439.1 | NP_001004439.1 | 8 | 950 | ||
XM_005254228.3 | XP_005254285.1 | 6 | 1172 | |||
XM_011521363.2 | XP_011519665.1 | 6 | 902 | |||
GenBank mRNA | AF109681.1 | - | 8 | 935 | ||
AF137378.2 | - | 8 | 953 | |||
AK095863.1 | - | 9 | 962 | |||
AK292445.1 | - | 8 | 959 | |||
AK304778.1 | - | 8 | 960 |