Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004439.1 | NP_001004439.1 | 30 | 3982 | ||
XM_005254228.3 | XP_005254285.1 | 28 | 4204 | |||
XM_011521363.2 | XP_011519665.1 | 28 | 3934 | |||
GenBank mRNA | AF109681.1 | - | 30 | 3970 | ||
AK304778.1 | - | 16 | 2353 | |||
AL359064.1 | - | 8 | 1038 |