Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142434.1 | NP_001135906.1 | 3, 4 | 797 | ||
NM_012215.3 | NP_036347.1 | 3, 4 | 797 | |||
GenBank mRNA | AB014579.2 | - | 3, 4 | 359 | ||
AF036144.2 | - | 3, 4 | 749 | |||
AF307332.1 | - | 3, 4 | 749 | |||
BC039583.2 | - | 3, 4 | 756 | 5558183 | ||
BC047877.1 | - | 3, 4 | 752 |