Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017524.2 | NP_001017524.1 | 7 | 1196 | ||
NM_012239.5 | NP_036371.1 | 7 | 1342 | |||
XM_005252835.1 | XP_005252892.1 | 7 | 1396 | |||
XM_011519956.1 | XP_011518258.1 | 7 | 2188 | |||
XM_011519957.1 | XP_011518259.1 | 7 | 2017 | |||
XM_017017428.1 | XP_016872917.1 | 7 | 1242 | |||
XM_017017429.1 | XP_016872918.1 | 7 | 1416 | |||
XM_017017430.1 | XP_016872919.1 | 7 | 1356 | |||
XM_017017431.1 | XP_016872920.1 | 7 | 1141 | |||
GenBank mRNA | AF083108.2 | - | 7 | 1410 | ||
AK074992.1 | - | 5 | 2182 | |||
AK298572.1 | - | 7 | 1091 | |||
AK299009.1 | - | 6 | 1170 | |||
AK299438.1 | - | 7 | 1174 | |||
AK301558.1 | - | 6 | 1196 | |||
AK301979.1 | - | 7 | 1384 | |||
AL137276.1 | - | 3 | 465 | |||
BC001042.2 | - | 7 | 1325 | 2823471 | ||
DR762907.1 | - | 1 | 56 |