Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012248.3 | NP_036380.2 | 1 | 814 | ||
GenBank mRNA | AK092003.1 | - | 3 | 566 | ||
AK093889.1 | - | 2 | 571 | |||
AK303275.1 | - | 2 | 626 | |||
BC002381.2 | - | 1 | 698 | 2961275 | ||
BC016643.1 | - | 1 | 687 | 3923979 | ||
U43286.1 | - | 1 | 704 |