Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012287.5 | NP_036419.3 | 23 | 2502 | ||
XM_006713557.3 | XP_006713620.1 | 24 | 2523 | |||
XM_011512602.2 | XP_011510904.1 | 25 | 2628 | |||
XM_011512603.2 | XP_011510905.1 | 24 | 2607 | |||
XM_011512604.2 | XP_011510906.1 | 26 | 2787 | |||
XM_011512605.2 | XP_011510907.1 | 25 | 2549 | |||
XM_011512606.2 | XP_011510908.1 | 25 | 2530 | |||
XM_017006046.1 | XP_016861535.1 | 26 | 2736 | |||
XM_017006047.1 | XP_016861536.1 | 25 | 2715 | |||
XM_017006048.1 | XP_016861537.1 | 25 | 2631 | |||
XM_017006049.1 | XP_016861538.1 | 24 | 2610 | |||
GenBank mRNA | AJ238248.1 | - | 23 | 2321 | ||
AK290369.1 | - | 23 | 2434 | |||
BC031005.1 | - | 6 | 530 | |||
BC060767.1 | - | 23 | 2437 | 30343546 | ||
D26069.3 | - | 23 | 2271 |