Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L3 SIRNA Assays ›
Gene Symbol:
EPB41L3
Gene Name:
erythrocyte membrane protein band 4.1 like 3
Gene Aliases:
4.1B, DAL-1, DAL1
Chromosome Location:
Chr.18: 5392381 - 5630666 on Build GRCh38
UniGene ID:
Hs.213394
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136489
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281533.1
NP_001268462.1
3
464
NM_001281534.1
NP_001268463.1
3
645
NM_001281535.1
NP_001268464.1
5
446
NM_012307.3
NP_036439.2
3
645
XM_011525623.2
XP_011523925.1
3
645
XM_011525626.2
XP_011523928.1
2
247
XM_011525628.2
XP_011523930.1
3
645
XM_011525632.2
XP_011523934.1
2
247
XM_011525634.2
XP_011523936.1
2
247
XM_011525637.2
XP_011523939.1
2
247
XM_017025618.1
XP_016881107.1
2
247
XM_017025619.1
XP_016881108.1
2
247
XM_017025620.1
XP_016881109.1
2
247
XM_017025621.1
XP_016881110.1
2
247
XM_017025622.1
XP_016881111.1
3
645
XM_017025623.1
XP_016881112.1
2
247
XM_017025624.1
XP_016881113.1
2
247
XM_017025625.1
XP_016881114.1
3
413
XM_017025626.1
XP_016881115.1
2
247
XM_017025627.1
XP_016881116.1
3
645
XM_017025628.1
XP_016881117.1
3
645
XM_017025629.1
XP_016881118.1
3
645
XM_017025630.1
XP_016881119.1
3
413
XM_017025631.1
XP_016881120.1
2
247
XM_017025632.1
XP_016881121.1
3
645
XM_017025633.1
XP_016881122.1
3
413
XM_017025634.1
XP_016881123.1
2
247
XM_017025635.1
XP_016881124.1
2
247
XM_017025636.1
XP_016881125.1
3
645
XM_017025637.1
XP_016881126.1
3
645
XM_017025638.1
XP_016881127.1
2
247
XM_017025639.1
XP_016881128.1
3
645
XM_017025640.1
XP_016881129.1
3
645
XM_017025641.1
XP_016881130.1
2
247
XM_017025642.1
XP_016881131.1
3
645
XM_017025643.1
XP_016881132.1
3
645
XM_017025644.1
XP_016881133.1
3
645
XM_017025645.1
XP_016881134.1
3
504
XM_017025646.1
XP_016881135.1
3
645
XM_017025647.1
XP_016881136.1
3
504
XM_017025648.1
XP_016881137.1
3
504
XM_017025649.1
XP_016881138.1
3
504
XM_017025650.1
XP_016881139.1
3
504
XM_017025651.1
XP_016881140.1
3
504
XM_017025652.1
XP_016881141.1
3
504
XM_017025653.1
XP_016881142.1
3
504
XM_017025654.1
XP_016881143.1
3
504
XM_017025655.1
XP_016881144.1
3
504
XM_017025656.1
XP_016881145.1
3
504
XM_017025657.1
XP_016881146.1
3
504
XM_017025658.1
XP_016881147.1
3
504
XM_017025659.1
XP_016881148.1
3
504
XM_017025660.1
XP_016881149.1
3
504
GenBank mRNA
AB023204.1
-
3
322
AF069072.1
-
3
302
AK128531.1
-
4
313
AK292583.1
-
5
446
AK297406.1
-
3
464
AK307904.1
-
3
566
BC006141.1
-
3
616
3936453
BC008377.1
-
3
481
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
protein localization to paranode region of axon
apoptotic process
cytoskeletal anchoring at plasma membrane
biological_process
regulation of cell shape
cortical cytoskeleton organization
cortical actin cytoskeleton organization
paranodal junction assembly
myelin maintenance
neuron projection morphogenesis
protein localization to juxtaparanode region of axon
protein localization to plasma membrane
actin binding
structural constituent of cytoskeleton
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0