Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L3 SIRNA Assays ›
Gene Symbol:
EPB41L3
Gene Name:
erythrocyte membrane protein band 4.1 like 3
Gene Aliases:
4.1B, DAL-1, DAL1
Chromosome Location:
Chr.18: 5392381 - 5630666 on Build GRCh38
UniGene ID:
Hs.213394
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136491
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281533.1
NP_001268462.1
21
3073
NM_001281534.1
NP_001268463.1
20
3212
NM_001281535.1
NP_001268464.1
22
3013
NM_012307.3
NP_036439.2
23
3878
XM_011525623.2
XP_011523925.1
23
3446
XM_011525626.2
XP_011523928.1
21
3012
XM_011525628.2
XP_011523930.1
22
3371
XM_011525632.2
XP_011523934.1
21
2925
XM_011525634.2
XP_011523936.1
20
2862
XM_011525637.2
XP_011523939.1
18
2760
XM_017025618.1
XP_016881107.1
20
2931
XM_017025619.1
XP_016881108.1
20
2895
XM_017025620.1
XP_016881109.1
20
2889
XM_017025621.1
XP_016881110.1
19
2883
XM_017025622.1
XP_016881111.1
23
3392
XM_017025623.1
XP_016881112.1
19
2856
XM_017025624.1
XP_016881113.1
20
2844
XM_017025625.1
XP_016881114.1
22
3139
XM_017025626.1
XP_016881115.1
19
2820
XM_017025627.1
XP_016881116.1
22
3365
XM_017025628.1
XP_016881117.1
21
3335
XM_017025629.1
XP_016881118.1
22
3317
XM_017025630.1
XP_016881119.1
22
3085
XM_017025631.1
XP_016881120.1
18
2766
XM_017025632.1
XP_016881121.1
21
3290
XM_017025633.1
XP_016881122.1
21
3058
XM_017025634.1
XP_016881123.1
19
2733
XM_017025635.1
XP_016881124.1
18
2745
XM_017025636.1
XP_016881125.1
21
3254
XM_017025637.1
XP_016881126.1
21
3248
XM_017025638.1
XP_016881127.1
18
2697
XM_017025639.1
XP_016881128.1
21
3236
XM_017025640.1
XP_016881129.1
20
3224
XM_017025641.1
XP_016881130.1
17
2679
XM_017025642.1
XP_016881131.1
21
3200
XM_017025643.1
XP_016881132.1
20
3167
XM_017025644.1
XP_016881133.1
20
3158
XM_017025645.1
XP_016881134.1
23
3305
XM_017025646.1
XP_016881135.1
19
3131
XM_017025647.1
XP_016881136.1
22
3269
XM_017025648.1
XP_016881137.1
22
3230
XM_017025649.1
XP_016881138.1
23
3251
XM_017025650.1
XP_016881139.1
22
3224
XM_017025651.1
XP_016881140.1
21
3194
XM_017025652.1
XP_016881141.1
22
3176
XM_017025653.1
XP_016881142.1
22
3170
XM_017025654.1
XP_016881143.1
21
3149
XM_017025655.1
XP_016881144.1
21
3140
XM_017025656.1
XP_016881145.1
21
3113
XM_017025657.1
XP_016881146.1
21
3107
XM_017025658.1
XP_016881147.1
21
3095
XM_017025659.1
XP_016881148.1
20
3071
XM_017025660.1
XP_016881149.1
21
3053
GenBank mRNA
AB023204.1
-
23
3555
AF069072.1
-
20
2866
AF515797.1
-
1
155
AK090875.1
-
1
1370
AK094952.1
-
10
1358
AK128531.1
-
22
2880
AK292583.1
-
23
3013
AK294690.1
-
11
1535
AK295453.1
-
12
1556
AK297406.1
-
21
3073
AK303679.1
-
12
1653
AL832598.2
-
5
4014
BC006141.1
-
21
3183
3936453
BC008377.1
-
9
1234
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
protein localization to paranode region of axon
apoptotic process
cytoskeletal anchoring at plasma membrane
biological_process
regulation of cell shape
cortical cytoskeleton organization
cortical actin cytoskeleton organization
paranodal junction assembly
myelin maintenance
neuron projection morphogenesis
protein localization to juxtaparanode region of axon
protein localization to plasma membrane
actin binding
structural constituent of cytoskeleton
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0