Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012308.2 | NP_036440.1 | 9 | 1685 | ||
NR_027473.1 | 9 | 1685 | ||||
XM_006718480.3 | XP_006718543.1 | 5 | 703 | |||
XM_011544860.2 | XP_011543162.1 | 9 | 1744 | |||
XM_011544861.1 | XP_011543163.1 | 4 | 397 | |||
XM_017017391.1 | XP_016872880.1 | 9 | 991 | |||
XM_017017392.1 | XP_016872881.1 | 8 | 802 | |||
GenBank mRNA | AB023221.2 | - | 9 | 1002 | ||
AK024505.1 | - | 9 | 927 | |||
BC047371.1 | - | 4 | 397 | 5270744 | ||
BC047486.1 | - | 9 | 1687 | 5296445 | ||
BC064360.1 | - | 9 | 1100 | 5534384 | ||
JQ710741.1 | - | 8 | 869 | |||
JQ710742.1 | - | 9 | 991 | |||
JQ710743.1 | - | 8 | 869 | |||
JQ710744.1 | - | 10 | 1038 |