Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256405.1 | NP_001243334.1 | 2, 3 | 420 | ||
NM_012308.2 | NP_036440.1 | 13, 14 | 2428 | |||
NR_027473.1 | 13, 14 | 2428 | ||||
XM_006718480.3 | XP_006718543.1 | 9, 10 | 1446 | |||
XM_011544861.1 | XP_011543163.1 | 8, 9 | 1140 | |||
XM_017017391.1 | XP_016872880.1 | 13, 14 | 1734 | |||
XM_017017392.1 | XP_016872881.1 | 12, 13 | 1545 | |||
GenBank mRNA | AB023221.2 | - | 13, 14 | 1745 | ||
AB490246.1 | - | 2, 3 | 408 | |||
AK024505.1 | - | 13, 14 | 1669 | |||
AK299725.1 | - | 2, 3 | 351 | |||
BC047371.1 | - | 8, 9 | 1140 | 5270744 | ||
BC047486.1 | - | 13, 14 | 2430 | 5296445 | ||
BC064360.1 | - | 13, 14 | 1843 | 5534384 | ||
DR001963.1 | - | 2, 3 | 427 | |||
JQ710741.1 | - | 12, 13 | 1612 | |||
JQ710742.1 | - | 13, 14 | 1734 | |||
JQ710743.1 | - | 12, 13 | 1612 | |||
JQ710744.1 | - | 14, 15 | 1648 |