Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012325.2 | NP_036457.1 | 5, 6 | 741 | ||
XM_011528696.2 | XP_011526998.1 | 5, 6 | 743 | |||
GenBank mRNA | AK297273.1 | - | 5, 6 | 590 | ||
AK310817.1 | - | 2, 3 | 146 | |||
AK312590.1 | - | 5, 6 | 658 | |||
BC040410.1 | - | 5, 6 | 668 | |||
BC106068.1 | - | 5, 6 | 671 | 6527006 | ||
BC109281.3 | - | 5, 6 | 665 | 40031641 | ||
U24166.1 | - | 5, 6 | 666 |