Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012329.2 | NP_036461.2 | 7 | 1546 | ||
XM_006721795.2 | XP_006721858.1 | 8 | 1652 | |||
GenBank mRNA | AY424289.1 | - | 7 | 1526 | ||
BC026324.1 | - | 7 | 1518 | 4794375 | ||
X85750.1 | - | 7 | 1343 |