Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5K1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195733.1 | NP_001182662.1 | 17 | 4255 | ||
NM_012398.2 | NP_036530.1 | 18 | 4339 | |||
XM_017026540.1 | XP_016882029.1 | 17 | 4313 | |||
GenBank mRNA | AB011161.1 | - | 18 | 4307 | ||
BC011138.1 | - | 1 | 1294 |