Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204204.2 | NP_001191133.1 | 10 | 1271 | ||
NM_001291932.1 | NP_001278861.1 | 11 | 1444 | |||
NM_012429.4 | NP_036561.1 | 12 | 1520 | |||
GenBank mRNA | AB033012.2 | - | 12 | 1419 | ||
AK056163.1 | - | 6 | 889 | |||
AK091402.1 | - | 10 | 1307 | |||
AK092576.1 | - | 1 | 909 | |||
AK296540.1 | - | 11 | 1334 | |||
AK303751.1 | - | 10 | 1167 | |||
AL096881.1 | - | 12 | 1517 |