Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204204.2 | NP_001191133.1 | 10 | 2552 | ||
NM_001291932.1 | NP_001278861.1 | 11 | 2725 | |||
NM_012429.4 | NP_036561.1 | 12 | 2801 | |||
GenBank mRNA | AB033012.2 | - | 12 | 2700 | ||
AK056163.1 | - | 6 | 2170 | |||
AK091402.1 | - | 10 | 2589 | |||
AL096881.1 | - | 12 | 2798 | |||
AY203959.1 | - | 1 | 1114 |