Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF049907.1 | - | 3 | 1367 | ||
AK026123.1 | - | 3 | 1333 | |||
BC008828.2 | - | 3 | 1311 | 4123095 | ||
BT007194.1 | - | 1 | 1114 |