Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SACM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319072.1 | NP_001306001.1 | 2 | 312 | ||
NM_001319073.1 | NP_001306002.1 | 3 | 352 | |||
NM_014016.4 | NP_054735.3 | 2 | 312 | |||
XM_011533500.2 | XP_011531802.1 | 3 | 231 | |||
GenBank mRNA | AB020658.1 | - | 2 | 156 | ||
AK291142.1 | - | 2 | 196 | |||
AK296421.1 | - | 2 | 94 | |||
AK301248.1 | - | 3 | 192 | |||
AL136831.1 | - | 2 | 102 | |||
BC016559.1 | - | 2 | 108 | 3049075 | ||
CR749564.1 | - | 2 | 1258 | |||
DC317560.1 | - | 2 | 291 | |||
DC322649.1 | - | 3 | 352 |