Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318151.1 | NP_001305080.1 | 3 | 520 | ||
NM_001318152.1 | NP_001305081.1 | 2 | 490 | |||
NM_003776.3 | NP_003767.2 | 3 | 870 | |||
GenBank mRNA | AF034091.1 | - | 3 | 229 | ||
AJ295637.1 | - | 3 | 227 | |||
AK057708.1 | - | 1 | 2256 | |||
AK123768.1 | - | 3 | 870 | |||
BC009707.1 | - | 3 | 259 | 3876531 | ||
BP309678.1 | - | 2 | 490 | |||
BX382916.2 | - | 3 | 444 | |||
CR456532.1 | - | 3 | 222 |