Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014255.5 | NP_055070.1 | 4 | 914 | ||
GenBank mRNA | AB015631.1 | - | 4 | 509 | ||
AK312921.1 | - | 4 | 611 | |||
AY032624.1 | - | 4 | 783 | |||
AY359102.1 | - | 4 | 907 | |||
BC065015.1 | - | 4 | 549 | 6169229 |