Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014258.3 | NP_055073.2 | 28 | 2874 | ||
XM_011528487.2 | XP_011526789.1 | 28 | 2910 | |||
XM_011528488.2 | XP_011526790.1 | 28 | 2862 | |||
XM_011528489.2 | XP_011526791.1 | 28 | 2905 | |||
XM_011528490.2 | XP_011526792.1 | 28 | 2886 | |||
XM_011528493.2 | XP_011526795.1 | 27 | 2825 | |||
XM_017027586.1 | XP_016883075.1 | 27 | 3106 | |||
XM_017027587.1 | XP_016883076.1 | 27 | 3103 | |||
XM_017027588.1 | XP_016883077.1 | 27 | 3107 | |||
XM_017027589.1 | XP_016883078.1 | 27 | 3107 | |||
XM_017027590.1 | XP_016883079.1 | 27 | 2799 | |||
XM_017027591.1 | XP_016883080.1 | 19 | 2205 | |||
GenBank mRNA | BC040566.1 | - | 28 | 2910 | ||
BC132864.1 | - | 27 | 2743 | 40146886 | ||
BC132870.1 | - | 27 | 2745 | 40146892 | ||
BX648091.1 | - | 28 | 2827 | |||
BX649108.1 | - | 28 | 2852 | |||
Y08982.1 | - | 26 | 2653 |