Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143826.2 | NP_001137298.1 | 3, 4 | 353 | ||
NM_001143827.2 | NP_001137299.1 | 3, 4 | 499 | |||
NM_001256420.1 | NP_001243349.1 | 1, 2 | 313 | |||
NM_014268.3 | NP_055083.1 | 2, 3 | 441 | |||
NR_046177.1 | 2, 3 | 256 | ||||
GenBank mRNA | AB016823.1 | - | 2, 3 | 376 | ||
AK090945.1 | - | 1, 2 | 269 | |||
AK294833.1 | - | 3, 4 | 443 | |||
AK296251.1 | - | 2, 3 | 256 | |||
AK315766.1 | - | 2, 3 | 394 | |||
BC007318.2 | - | 2, 3 | 384 | 2989643 | ||
BT020086.1 | - | 2, 3 | 261 | |||
CR536545.1 | - | 2, 3 | 261 | |||
DC335558.1 | - | 3, 4 | 501 | |||
DC341673.1 | - | 3, 4 | 353 | |||
DC354326.1 | - | 2, 3 | 441 | |||
X94232.1 | - | 2, 3 | 401 |