Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014280.2 | NP_055095.2 | 5 | 417 | ||
GenBank mRNA | AF083190.1 | - | 5 | 424 | ||
AF161433.1 | - | 2 | 101 | |||
AK300797.1 | - | 5 | 406 | |||
BC033159.1 | - | 5 | 402 | 4330676 | ||
BX538122.1 | - | 6 | 534 | |||
CR456900.1 | - | 5 | 417 | |||
DB294321.1 | - | 5 | 417 |