Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317804.1 | NP_001304733.1 | 12 | 2260 | ||
NM_001317805.1 | NP_001304734.1 | 11 | 2775 | |||
NM_014294.5 | NP_055109.1 | 11 | 2343 | |||
XM_017013266.1 | XP_016868755.1 | 10 | 2215 | |||
XM_017013267.1 | XP_016868756.1 | 10 | 2559 | |||
GenBank mRNA | AF274933.1 | - | 1 | 462 | ||
BC000687.2 | - | 11 | 2165 | 3347823 | ||
BC037738.1 | - | 11 | 2255 | 5261490 | ||
BQ233660.1 | - | 1 | 578 |