Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145001.2 | NP_001138473.1 | 11 | 2332 | ||
NM_001166167.1 | NP_001159639.1 | 10 | 2114 | |||
NM_001166168.1 | NP_001159640.1 | 10 | 2109 | |||
NM_001166169.1 | NP_001159641.1 | 10 | 2118 | |||
NM_001166170.1 | NP_001159642.1 | 10 | 2116 | |||
NM_001166171.1 | NP_001159643.1 | 11 | 2281 | |||
NM_014397.5 | NP_055212.2 | 10 | 2167 | |||
XM_005251664.1 | XP_005251721.1 | 10 | 2173 | |||
XM_006716936.2 | XP_006716999.1 | 10 | 2178 | |||
XM_017014217.1 | XP_016869706.1 | 11 | 2341 | |||
GenBank mRNA | AJ420460.1 | - | 1 | 372 | ||
BC000101.2 | - | 10 | 2113 | 3507262 | ||
BC012761.2 | - | 10 | 2114 | 3640103 |