Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM120A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286722.1 | NP_001273651.1 | 5 | 1216 | ||
NM_001286723.1 | NP_001273652.1 | 5 | 1216 | |||
NM_001286724.1 | NP_001273653.1 | 5 | 1216 | |||
NM_014612.4 | NP_055427.2 | 5 | 1216 | |||
XM_005251842.4 | XP_005251899.1 | 5 | 1168 | |||
XM_011518412.2 | XP_011516714.1 | 5 | 1033 | |||
XM_011518413.2 | XP_011516715.1 | 5 | 1033 | |||
XM_011518414.2 | XP_011516716.1 | 5 | 1033 | |||
XM_011518416.2 | XP_011516718.1 | 5 | 1034 | |||
GenBank mRNA | AF214737.1 | - | 5 | 1021 | ||
AF214738.1 | - | 5 | 1021 | |||
BC007879.2 | - | 4 | 528 | |||
BC098584.1 | - | 5 | 1192 | 30531603 | ||
BC111736.1 | - | 5 | 1014 | 40036337 | ||
CF135376.1 | - | 4 | 398 | |||
D80005.2 | - | 5 | 977 |