Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014891.6 | NP_055706.1 | 5, 6 | 629 | ||
GenBank mRNA | BC000684.2 | - | 5, 6 | 541 | 3347335 | |
BC007873.2 | - | 5, 6 | 505 | 4303959 | ||
BC071993.1 | - | 5, 6 | 539 | |||
U41745.1 | - | 5, 6 | 516 |