Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014891.6 | NP_055706.1 | 6 | 1272 | ||
GenBank mRNA | BC000684.2 | - | 6 | 1184 | 3347335 | |
BC007873.2 | - | 6 | 1148 | 4303959 | ||
BC071993.1 | - | 6 | 1182 | |||
BQ215882.1 | - | 1 | 46 | |||
CB852527.1 | - | 1 | 299 |