Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278458.1 | NP_001265387.1 | 9 | 1128 | ||
NM_001278460.1 | NP_001265389.1 | 7 | 1068 | |||
NM_001278461.1 | NP_001265390.1 | 6 | 1028 | |||
NM_014900.4 | NP_055715.3 | 6 | 1028 | |||
GenBank mRNA | AB023194.1 | - | 6 | 1022 | ||
AK002054.1 | - | 8 | 1009 | |||
AK225849.1 | - | 7 | 925 | |||
AK294937.1 | - | 9 | 1128 | |||
BC071588.1 | - | 6 | 1030 | 30345650 | ||
BX537877.1 | - | 2 | 2630 | |||
BX649112.1 | - | 11 |