Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014908.3 | NP_055723.1 | 1 | 548 | ||
GenBank mRNA | AB029017.1 | - | 1 | 434 | ||
AK225402.1 | - | 1 | 421 | |||
AK300180.1 | - | 1 | 401 | |||
AY358759.1 | - | 1 | 403 | |||
BC035556.1 | - | 1 | 401 | 5224439 | ||
BF056649.1 | - | 1 | 548 | |||
KU178553.1 | - | 1 | 118 |