Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014912.4 | NP_055727.3 | 3 | 1335 | ||
XM_006717714.3 | XP_006717777.1 | 3 | 1253 | |||
XM_006717715.3 | XP_006717778.1 | 3 | 1335 | |||
XM_011539514.2 | XP_011537816.1 | 3 | 1335 | |||
XM_011539518.2 | XP_011537820.1 | 3 | 1335 | |||
XM_011539519.2 | XP_011537821.1 | 3 | 1335 | |||
XM_011539522.2 | XP_011537824.1 | 3 | 1335 | |||
GenBank mRNA | AK127060.1 | - | 2 | 256 | ||
BC036444.2 | - | 3 | 1304 | 4838004 |