Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178137.1 | NP_001171608.1 | 6 | 1476 | ||
NM_014912.4 | NP_055727.3 | 6 | 1600 | |||
XM_005269630.4 | XP_005269687.1 | 6 | 1558 | |||
XM_006717714.3 | XP_006717777.1 | 7 | 1569 | |||
XM_006717715.3 | XP_006717778.1 | 7 | 1651 | |||
XM_011539514.2 | XP_011537816.1 | 6 | 1627 | |||
XM_011539515.2 | XP_011537817.1 | 7 | 1582 | |||
XM_011539517.2 | XP_011537819.1 | 5 | 1501 | |||
XM_011539518.2 | XP_011537820.1 | 7 | 1651 | |||
XM_017015927.1 | XP_016871416.1 | 3 | 364 | |||
GenBank mRNA | AB023157.1 | - | 6 | 1443 | ||
AK127060.1 | - | 5 | 548 | |||
AK302576.1 | - | 6 | 1545 | |||
BC036444.2 | - | 6 | 1569 | 4838004 |