Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003793.3 | NP_003784.2 | 3, 4 | 629 | ||
XM_011545328.2 | XP_011543630.1 | 3, 4 | 379 | |||
GenBank mRNA | AF071748.1 | - | 1, 2 | 193 | ||
AF071749.1 | - | 1, 2 | 95 | |||
AF088886.2 | - | 3, 4 | 629 | |||
AF136279.1 | - | 3, 4 | 628 | |||
AJ007331.1 | - | 3, 4 | 584 | |||
AK313657.1 | - | 3, 4 | 597 | |||
AL137742.1 | - | 1, 2 | 60 | |||
BC011682.2 | - | 3, 4 | 623 | 3535532 | ||
BC036451.1 | - | 3, 4 | 604 | 4830274 | ||
CR541680.1 | - | 1, 2 | 101 |