Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143854.1 | NP_001137326.1 | 8 | 879 | ||
NM_014954.3 | NP_055769.2 | 7 | 867 | |||
XM_011538048.1 | XP_011536350.1 | 8 | 683 | |||
GenBank mRNA | AB023202.1 | - | 8 | 800 | ||
AK295696.1 | - | 8 | 833 | |||
AK296296.1 | - | 6 | 418 | |||
AK316112.1 | - | 8 | 714 | |||
BC002605.2 | - | 6 | 605 | |||
BC009843.2 | - | 6 | 528 | |||
BC017259.2 | - | 7 | 674 | 3510158 | ||
BX647281.1 | - | 1 | 310 |