Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK38L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015000.3 | NP_055815.1 | 13 | 1388 | ||
XM_005253342.3 | XP_005253399.1 | 12 | 1267 | |||
XM_006719058.3 | XP_006719121.1 | 14 | 1369 | |||
XM_006719059.3 | XP_006719122.1 | 14 | 1348 | |||
XM_011520613.2 | XP_011518915.1 | 12 | 1082 | |||
XM_017019036.1 | XP_016874525.1 | 13 | 1344 | |||
GenBank mRNA | AB023182.2 | - | 13 | 1473 | ||
AK291055.1 | - | 13 | 1278 | |||
AK303373.1 | - | 3 | 548 | |||
AK304755.1 | - | 11 | 972 | |||
AK304862.1 | - | 7 | 684 | |||
BC028603.1 | - | 13 | 1362 | 4823429 | ||
BX390969.1 | - | 6 | 473 |