Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015024.4 | NP_055839.3 | 3 | 317 | ||
XM_017013244.1 | XP_016868733.1 | 3 | 544 | |||
XM_017013245.1 | XP_016868734.1 | 3 | 311 | |||
XM_017013246.1 | XP_016868735.1 | 3 | 540 | |||
XM_017013247.1 | XP_016868736.1 | 3 | 704 | |||
GenBank mRNA | AB018288.1 | - | 3 | 227 | ||
AF064729.1 | - | 3 | 227 | |||
AK291472.1 | - | 3 | 232 | |||
AK311616.1 | - | 3 | 410 | |||
AL597247.1 | - | 3 | 273 | |||
BC030785.1 | - | 3 | 316 | 5269230 | ||
DB460798.1 | - | 3 | 319 | |||
DC332379.1 | - | 3 | 239 |